Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AIM23
Entrez Gene ID
853310
Gene Name
Gene Aliases
MIF3, YJL131C
Chromosome Location
Chr.X: 164278 - 165348 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181564.1 NP_012404.1 1 - 1 953 94

View Full Product Details