Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IRC19
Entrez Gene ID
850626
Gene Name
Gene Aliases
RRG4, YLL033W
Chromosome Location
Chr.XII: 73409 - 74101 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181853.1 NP_013067.1 1 - 1 349 83

View Full Product Details