Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UTP13
Entrez Gene ID
850919
Gene Name
Gene Aliases
YLR222C
Chromosome Location
Chr.XII: 579318 - 581771 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182109.1 NP_013323.1 1 - 1 2347 70

View Full Product Details