Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VID22
Entrez Gene ID
851088
Gene Name
Gene Aliases
YLR373C
Chromosome Location
Chr.XII: 868662 - 871367 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182262.1 NP_013477.1 1 - 1 2646 108

View Full Product Details