Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TVP18
Entrez Gene ID
855093
Gene Name
Gene Aliases
YMR071C
Chromosome Location
Chr.XIII: 410762 - 411265 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182569.1 NP_013787.1 1 - 1 455 99

View Full Product Details