Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPI15
Entrez Gene ID
855690
Gene Name
Gene Aliases
YNL038W
Chromosome Location
Chr.XIV: 557020 - 557783 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182877.1 NP_014360.2 1 - 2 583 95

View Full Product Details