Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTC5
Entrez Gene ID
854257
Gene Name
Gene Aliases
PPP1, YOR090C
Chromosome Location
Chr.XV: 491123 - 492841 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183509.1 NP_014733.1 1 - 1 1642 77

View Full Product Details