Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRM1
Entrez Gene ID
854376
Gene Name
Gene Aliases
PET56, YOR201C
Chromosome Location
Chr.XV: 720470 - 721708 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183620.3 NP_014844.3 1 - 1 1150 75

View Full Product Details