Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VTS1
Entrez Gene ID
854541
Gene Name
Gene Aliases
YOR359W
Chromosome Location
Chr.XV: 1011189 - 1012760 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183779.3 NP_015004.3 1 - 1 1445 95

View Full Product Details