Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRM44
Entrez Gene ID
856077
Gene Name
Gene Aliases
YPL030W
Chromosome Location
Chr.XVI: 493543 - 495246 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183844.1 NP_015295.1 1 - 1 1557 101

View Full Product Details