Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LEE1
Entrez Gene ID
856053
Gene Name
Gene Aliases
YPL054W
Chromosome Location
Chr.XVI: 455759 - 456664 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183868.1 NP_015271.1 1 - 1 748 71

View Full Product Details