Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NHP6A
- Entrez Gene ID
-
856165
- Gene Name
- Gene Aliases
-
YPR052C
- Chromosome Location
-
Chr.XVI: 665693 - 665974 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001184149.1 |
NP_015377.1 |
1 - 1 |
55 |
80 |
- Gene Symbol
- 856166
- Entrez Gene ID
-
856166
- Gene Name
- Gene Aliases
-
YPR053C
- Chromosome Location
-
Chr.XVI: 665788 - 666243 on Build R64
View Full Product Details