Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
TNNI2
Entrez Gene ID
606751
Gene Name
troponin I2, fast skeletal type
Gene Aliases
TnI-F4
Chromosome Location
Chr.2: 1251566 - 1253883 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032359.1 NP_001027530.1 5 - 5 151 68
GenBank mRNA AK347244.1 - 2 - 2 65 68
DQ091303.1 - 5 - 5 151 68
EU751622.1 - 2 - 2 106 68
FJ719788.1 - 5 - 5 190 68

View Full Product Details