Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
NME2
Entrez Gene ID
733683
Gene Name
NME/NM23 nucleoside diphosphate kinase 2
Gene Aliases
-
Chromosome Location
Chr.12: 27439529 - 27445025 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044610.2 NP_001038075.1 4 - 4 410 88
GenBank mRNA AK350066.1 - 4 - 4 412 88
DQ372081.1 - 3 - 3 254 88

View Full Product Details