Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HRH4
Entrez Gene ID
397224
Gene Name
histamine receptor H4
Gene Aliases
-
Chromosome Location
Chr.6: 109302264 - 109318607 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214122.1 NP_999287.1 3 - 3 1148 121
GenBank mRNA AB053300.1 - 3 - 3 1148 121

View Full Product Details