Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRAT
Entrez Gene ID
100127351
Gene Name
carnitine O-acetyltransferase
Gene Aliases
-
Chromosome Location
Chr.1: 269387430 - 269401334 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113047.1 NP_001106518.1 12 - 13 1645 58
XM_005660485.3 XP_005660542.2 - 1876 58
XM_021077790.1 XP_020933449.1 - 1654 58
XM_021078296.1 XP_020933955.1 - 2350 58
GenBank mRNA AM919499.1 - 12 - 13 1645 58

View Full Product Details