Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HADHB
Entrez Gene ID
397017
Gene Name
hydroxyacyl-CoA dehydrogenase trifunctional multienzyme complex subunit beta
Gene Aliases
LCTHIO
Chromosome Location
Chr.3: 112712894 - 112753121 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001348972.1 NP_001335901.1 14 - 15 1361 92
GenBank mRNA AF028007.1 - 11 - 12 1125 92
AK239646.1 - 14 - 15 1308 92
AK389670.1 - 14 - 15 1329 92
AK393956.1 - 14 - 15 1297 92
AK400872.1 - 14 - 15 1282 92

View Full Product Details