Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
LPL
Entrez Gene ID
397537
Gene Name
lipoprotein lipase
Gene Aliases
-
Chromosome Location
Chr.14: 4104761 - 4134964 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214286.1 NP_999451.1 9 - 10 1578 66
GenBank mRNA AK344311.1 - 9 - 10 1617 66
AY559452.1 - 9 - 10 1557 66
AY559453.1 - 9 - 10 1557 66
AY559454.1 - 9 - 10 1557 66
AY686761.1 - 9 - 10 1557 66
X62984.1 - - 1595 66

View Full Product Details