Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MCL1
Entrez Gene ID
397648
Gene Name
MCL1 apoptosis regulator, BCL2 family member
Gene Aliases
-
Chromosome Location
Chr.4: 98562092 - 98567014 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001348806.1 NP_001335735.1 2 - 3 1078 81
GenBank mRNA AJ307006.1 - 2 - 3 404 81
AK232161.1 - 2 - 3 1016 81
AK389656.1 - 1 - 2 312 81
AK392300.1 - 2 - 3 1079 81

View Full Product Details