Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSMB6
Entrez Gene ID
497065
Gene Name
proteasome 20S subunit beta 6
Gene Aliases
-
Chromosome Location
Chr.12: 52096009 - 52098164 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144926.2 NP_001138398.2 3 - 3 249 84
XM_021066255.1 XP_020921914.1 - 272 84
GenBank mRNA AK234635.1 - 3 - 3 248 84
AK234947.1 - 3 - 3 230 84
AK391728.1 - 3 - 3 250 84
AK400437.1 - 3 - 3 339 84
AY609990.1 - 3 - 3 236 84
CJ013147.1 - 3 - 3 249 84
HQ288834.1 - 3 - 3 234 84

View Full Product Details