Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPATA7
Entrez Gene ID
100153278
Gene Name
spermatogenesis associated 7
Gene Aliases
-
Chromosome Location
Chr.7: 110309414 - 110368370 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177908.1 NP_001171379.1 8 - 9 1104 70
XM_013978348.2 XP_013833802.1 - 1056 70
XM_013978349.2 XP_013833803.1 - 968 70
GenBank mRNA AK347650.1 - 8 - 9 1104 70
GU475000.1 - 8 - 9 1104 70

View Full Product Details