Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FDPS
Entrez Gene ID
100153932
Gene Name
farnesyl diphosphate synthase
Gene Aliases
-
Chromosome Location
Chr.4: 94500140 - 94514820 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185131.1 NP_001172060.1 9 - 10 1068 57
XM_005663310.3 XP_005663367.1 - 994 57
XM_021088556.1 XP_020944215.1 - 1226 57
XM_021088557.1 XP_020944216.1 - 1133 57
XM_021088559.1 XP_020944218.1 - 1093 57
GenBank mRNA AB529527.1 - 9 - 10 1068 57
AK236486.1 - 10 - 11 1289 57
AK351226.1 - 9 - 10 910 57
AK392898.1 - 9 - 10 995 57
AK398138.1 - 9 - 10 995 57
AK398787.1 - 10 - 11 1127 57
AY609787.1 - 9 - 10 1313 57

View Full Product Details