Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLAC9
Entrez Gene ID
100155282
Gene Name
placenta associated 9
Gene Aliases
-
Chromosome Location
Chr.14: 81890265 - 81901909 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190209.1 NP_001177138.1 3 - 4 425 56
GenBank mRNA AK235203.1 - 3 - 4 406 56
BP173023.1 - 3 - 4 333 56
CV868497.1 - 3 - 4 425 56

View Full Product Details