Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMIM7
Entrez Gene ID
100512765
Gene Name
small integral membrane protein 7
Gene Aliases
C19orf42, C2H19orf42
Chromosome Location
Chr.2: 60961532 - 60970775 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244720.1 NP_001231649.1 3 - 4 158 101
GenBank mRNA AK237926.1 - 2 - 3 297 101
AK239795.1 - 3 - 4 162 101
AK393070.1 - 3 - 4 158 101
AK393503.1 - - 158 101
AK399538.1 - 4 - 5 379 101

View Full Product Details