Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBCB
Entrez Gene ID
100522808
Gene Name
tubulin folding cofactor B
Gene Aliases
CKAP1
Chromosome Location
Chr.6: 45482542 - 45492627 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205403.1 NP_001192332.1 3 - 4 430 65
GenBank mRNA AK240439.1 - 3 - 4 422 65
AK352168.1 - 3 - 4 403 65
CJ016107.1 - 3 - 4 430 65

View Full Product Details