Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INIP
Entrez Gene ID
100513906
Gene Name
INTS3 and NABP interacting protein
Gene Aliases
C1H9orf80, C9orf80
Chromosome Location
Chr.1: 253375193 - 253393886 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244886.1 NP_001231815.1 4 - 5 406 85
XM_005660326.3 XP_005660383.1 - 490 85
XM_021071891.1 XP_020927550.1 - 360 85
GenBank mRNA AK232928.1 - 4 - 5 406 85

View Full Product Details