Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMOD4
Entrez Gene ID
100151861
Gene Name
tropomodulin 4
Gene Aliases
-
Chromosome Location
Chr.4: 98074380 - 98079346 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243294.1 NP_001230223.1 4 - 5 544 67
XM_005663458.3 XP_005663515.1 - 642 67
XM_013997159.2 XP_013852613.1 - 432 67
XM_021088491.1 XP_020944150.1 - 411 67
GenBank mRNA AK347176.1 - 3 - 4 734 67
EU649705.1 - 4 - 5 513 67

View Full Product Details