Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SFRP5
Entrez Gene ID
100153176
Gene Name
secreted frizzled related protein 5
Gene Aliases
-
Chromosome Location
Chr.14: 109120382 - 109125655 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243406.1 NP_001230335.1 1 - 2 578 101
GenBank mRNA AK350830.1 - 1 - 2 580 101
AK397123.1 - 1 - 2 609 101
DB781843.1 - 1 - 2 578 101
FD629736.1 - 1 - 2 162 101

View Full Product Details