Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HAUS2
Entrez Gene ID
100511852
Gene Name
HAUS augmin like complex subunit 2
Gene Aliases
-
Chromosome Location
Chr.1: 128820551 - 128839027 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243905.1 NP_001230834.1 4 - 5 410 80
GenBank mRNA AK232815.1 - 4 - 5 410 80
AY609754.1 - 4 - 5 409 80

View Full Product Details