Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IBA57
Entrez Gene ID
100513524
Gene Name
iron-sulfur cluster assembly factor IBA57
Gene Aliases
-
Chromosome Location
Chr.2: 51276352 - 51287572 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244830.1 NP_001231759.1 2 - 3 753 97
GenBank mRNA AK237626.1 - 2 - 3 753 97
AK396853.1 - 2 - 3 755 97

View Full Product Details