Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIR17
- Entrez Gene ID
-
100316572
- Gene Name
- microRNA mir-17
- Gene Aliases
-
ssc-mir-17
- Chromosome Location
-
Chr.11: 60734656 - 60734732 on Build Sscrofa11.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_035367.1 |
|
1 - 1 |
39 |
69 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
SLIT and NTRK like family member 5 |
SLITRK5 |
|
-2.9mb
|
Sense |
Chr. - 57792576 - 57800852 on Build Sscrofa11.1 |
Downstream |
microRNA mir-18a |
MIR18A |
MIR18, MIRN18, MIRN18A, ssc-mir-18, ssc-mir-18a |
59.0
|
Sense |
Chr. - 60734791 - 60734882 on Build Sscrofa11.1 |
View Full Product Details