Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GOX
Entrez Gene ID
100232932
Gene Name
glycolate oxidase-like
Gene Aliases
GSVIVT00028321001
Chromosome Location
Chr.10: 3369065 - 3374180 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY233203.1 - 3 - 4 286 77

View Full Product Details