Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sfrp5
Entrez Gene ID
448129
Gene Name
secreted frizzled-related protein 5
Gene Aliases
XsFRP5, frp5, sFRP-5, sarp3, sfrp5-a, sfrp5-b
Chromosome Location
Chr.7: 25554743 - 25597274 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004846.1 NP_001004846.1 2 - 3 897 96
GenBank mRNA BC074661.1 - 2 - 3 897 96

View Full Product Details