Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tvp23b
Entrez Gene ID
448668
Gene Name
trans-golgi network vesicle protein 23 homolog B (S. cerevisiae)
Gene Aliases
fam18b, fam18b1
Chromosome Location
Chr.10: 39233019 - 39237140 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005091.1 NP_001005091.1 2 - 3 133 79
GenBank mRNA BC077019.1 - 2 - 3 133 79

View Full Product Details