Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
c12orf57
Entrez Gene ID
394516
Gene Name
chromosome 12 open reading frame 57
Gene Aliases
-
Chromosome Location
Chr.7: 769590 - 774671 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203589.2 NP_988920.1 1 - 2 146 75
GenBank mRNA BC061620.1 - 1 - 2 77 75
BC075420.1 - 1 - 2 146 75
CR848390.2 - 1 - 2 106 75

View Full Product Details