Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sppl3
Entrez Gene ID
394539
Gene Name
signal peptide peptidase like 3
Gene Aliases
-
Chromosome Location
Chr.1: 143375172 - 143410325 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203611.1 NP_988942.1 4 - 5 450 84
GenBank mRNA BC061375.1 - 4 - 5 450 84

View Full Product Details