Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nucks1
Entrez Gene ID
394795
Gene Name
nuclear casein kinase and cyclin-dependent kinase substrate 1
Gene Aliases
JC7, nucks
Chromosome Location
Chr.2: 65087547 - 65101606 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203856.2 NP_989187.1 7 - 8 1198 79
XM_012958321.1 XP_012813775.1 7 - 8 1346 79
GenBank mRNA BC063335.1 - 7 - 8 924 79
CR760325.2 - 7 - 8 676 79

View Full Product Details