Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
rab6a
Entrez Gene ID
394940
Gene Name
RAB6A, member RAS oncogene family
Gene Aliases
rab6, rab6a', rab6b
Chromosome Location
Chr.Un|NW_016683507: 350791 - 372433 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203984.1 NP_989315.1 - 239 94
GenBank mRNA BC064230.1 - - 239 94

View Full Product Details