Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
wsb1
Entrez Gene ID
395021
Gene Name
WD repeat and SOCS box containing 1
Gene Aliases
WSB-1
Chromosome Location
Chr.2: 101207867 - 101221854 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204056.1 NP_989387.1 1 - 2 164 68
GenBank mRNA BC064858.1 - 1 - 2 164 68
CR942608.2 - 1 - 2 138 68

View Full Product Details