Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rraga
Entrez Gene ID
448744
Gene Name
Ras-related GTP binding A
Gene Aliases
-
Chromosome Location
Chr.9: 4642871 - 4648485 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006896.1 NP_001006897.1 3 - 4 191 75
GenBank mRNA BC074593.1 - 3 - 4 191 75
CR926416.2 - 2 - 3 154 75

View Full Product Details