Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dus4l
Entrez Gene ID
493309
Gene Name
dihydrouridine synthase 4-like
Gene Aliases
-
Chromosome Location
Chr.3: 66827840 - 66845221 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007928.1 NP_001007929.2 4 - 5 497 70
XM_018092045.1 XP_017947534.1 4 - 5 581 70
GenBank mRNA BC080375.1 - 4 - 5 497 70
CR761565.2 - 4 - 5 477 70

View Full Product Details