Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
esd
Entrez Gene ID
496692
Gene Name
esterase D
Gene Aliases
-
Chromosome Location
Chr.2: 131274374 - 131307081 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011247.1 NP_001011247.1 6 - 7 574 93
XM_012956849.2 XP_012812303.1 5 - 6 1128 93
XM_018091202.1 XP_017946691.1 6 - 7 1319 93
GenBank mRNA BC087823.1 - 6 - 7 574 93
CT030509.1 - 6 - 7 537 93

View Full Product Details