Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vsir
Entrez Gene ID
496981
Gene Name
V-set immunoregulatory receptor
Gene Aliases
c10orf54
Chromosome Location
Chr.7: 50831240 - 50869107 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011488.1 NP_001011488.1 4 - 5 704 94
GenBank mRNA BC088869.1 - 4 - 5 704 94

View Full Product Details