Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
syngr1
Entrez Gene ID
548949
Gene Name
synaptogyrin 1
Gene Aliases
P7D11
Chromosome Location
Chr.4: 110446858 - 110464765 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016195.2 NP_001016195.1 3 - 4 768 76
GenBank mRNA CR926351.2 - 3 - 4 768 76

View Full Product Details