Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
aimp1
Entrez Gene ID
549804
Gene Name
aminoacyl tRNA synthetase complex-interacting multifunctional protein 1
Gene Aliases
scye1
Chromosome Location
Chr.1: 62766750 - 62807911 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017050.2 NP_001017050.2 6 - 7 839 79
XM_012963225.1 XP_012818679.1 7 - 8 1110 79
GenBank mRNA BC075609.1 - 6 - 7 839 79

View Full Product Details