Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rab28
Entrez Gene ID
594989
Gene Name
RAB28, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.1: 17542811 - 17604233 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030430.1 NP_001025601.1 4 - 5 428 68
XM_018093837.1 XP_017949326.1 4 - 5 588 68
GenBank mRNA BC091050.1 - 4 - 5 428 68

View Full Product Details