Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pcyox1
Entrez Gene ID
549056
Gene Name
prenylcysteine oxidase 1
Gene Aliases
pcl1
Chromosome Location
Chr.3: 112125528 - 112133215 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016302.2 NP_001016302.1 4 - 5 719 77
GenBank mRNA BC170904.1 - 4 - 5 709 77
BC170906.1 - 4 - 5 709 77
CR761521.2 - 4 - 5 719 77

View Full Product Details