Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rab5if
Entrez Gene ID
549216
Gene Name
RAB5 interacting factor
Gene Aliases
RIP5, c20orf24
Chromosome Location
Chr.10: 32602101 - 32606318 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016462.2 NP_001016462.1 2 - 3 231 63
GenBank mRNA BC091613.1 - 2 - 3 253 63
CR760947.2 - 2 - 3 231 63

View Full Product Details