Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc39a11
Entrez Gene ID
549475
Gene Name
solute carrier family 39 member 11
Gene Aliases
-
Chromosome Location
Chr.10: 22684829 - 22878128 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016721.2 NP_001016721.1 2 - 3 173 64
XM_012966622.2 XP_012822076.1 3 - 4 257 64
GenBank mRNA CR760068.2 - 2 - 3 173 64

View Full Product Details