Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ppa1
Entrez Gene ID
549884
Gene Name
pyrophosphatase (inorganic) 1
Gene Aliases
pyp
Chromosome Location
Chr.1: 63154712 - 63173755 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017130.2 NP_001017130.1 3 - 4 320 87
GenBank mRNA BC082535.1 - 3 - 4 271 87
BC135836.1 - 3 - 4 246 87
BC168062.1 - 3 - 4 263 87
CR761093.2 - 3 - 4 320 87

View Full Product Details